missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxy-4-ethynylbenzene 98.0+%, TCI America™
$59.65 - $175.22
Chemical Identifiers
| CAS | 79887-14-2 |
|---|---|
| Molecular Formula | C10H10O |
| Molecular Weight (g/mol) | 146.19 |
| MDL Number | MFCD00173880 |
| InChI Key | FRGNOZUOTHMJSC-UHFFFAOYSA-N |
| Synonym | 4-Ethoxyphenylacetylene |
| PubChem CID | 2775124 |
| IUPAC Name | 1-ethoxy-4-ethynylbenzene |
| SMILES | CCOC1=CC=C(C=C1)C#C |
Chemical Identifiers
| 79887-14-2 | |
| 146.19 | |
| FRGNOZUOTHMJSC-UHFFFAOYSA-N | |
| 2775124 | |
| CCOC1=CC=C(C=C1)C#C |
| C10H10O | |
| MFCD00173880 | |
| 4-Ethoxyphenylacetylene | |
| 1-ethoxy-4-ethynylbenzene |
Specifications
| 79887-14-2 | |
| C10H10O | |
| 1 g | |
| FRGNOZUOTHMJSC-UHFFFAOYSA-N | |
| 1-ethoxy-4-ethynylbenzene | |
| 2775124 | |
| ≥98.0% (GC) | |
| 1-Ethoxy-4-ethynylbenzene |
| Yellow | |
| MFCD00173880 | |
| 4-Ethoxyphenylacetylene | |
| CCOC1=CC=C(C=C1)C#C | |
| 146.19 | |
| 146.19 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator