accessibility menu, dialog, popup

UserName

CH3OCH

CAS RN 79887-14-2

IUPAC Name: 1-ethoxy-4-ethynylbenzene
Synonyms: 1-ethoxy-4-ethynylbenzene
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: FRGNOZUOTHMJSC-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results