missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-4-ethynylbenzene 98.0+%, TCI America™

Catalog No. E10295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10295G 5 g
E10291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10295G Supplier TCI America Supplier No. E10295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79887-14-2
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00173880
InChI Key FRGNOZUOTHMJSC-UHFFFAOYSA-N
Synonym 4-Ethoxyphenylacetylene
PubChem CID 2775124
IUPAC Name 1-ethoxy-4-ethynylbenzene
SMILES CCOC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethoxy-4-ethynylbenzene
TSCA No
Recommended Storage Refrigerator