missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene 98.0+%, TCI America™
$95.06 - $462.29
Chemical Identifiers
| CAS | 174350-05-1 |
|---|---|
| Molecular Formula | C17H24F2O |
| Molecular Weight (g/mol) | 282.375 |
| MDL Number | MFCD11053400 |
| InChI Key | BOAHGIPRRKDVQY-UHFFFAOYSA-N |
| PubChem CID | 19103501 |
| IUPAC Name | 1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene |
| SMILES | CCCC1CCC(CC1)C2=C(C(=C(C=C2)OCC)F)F |
Chemical Identifiers
| 174350-05-1 | |
| 282.375 | |
| BOAHGIPRRKDVQY-UHFFFAOYSA-N | |
| 1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene |
| C17H24F2O | |
| MFCD11053400 | |
| 19103501 | |
| CCCC1CCC(CC1)C2=C(C(=C(C=C2)OCC)F)F |
Specifications
| 174350-05-1 | |
| C17H24F2O | |
| 1 g | |
| CCCC1CCC(CC1)C2=C(C(=C(C=C2)OCC)F)F | |
| 282.375 | |
| 282.37 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD11053400 | |
| BOAHGIPRRKDVQY-UHFFFAOYSA-N | |
| 1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene | |
| 19103501 | |
| ≥98.0% (GC) | |
| 1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene |
Safety and Handling
EINECSNumber : (4)-1953
TSCA : No