missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 97%, Thermo Scientific Chemicals

Catalog No. AC223450050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223450050 5 mL Glass bottle
AC223450250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223450050 Supplier Thermo Scientific Chemicals Supplier No. 223450050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 765-03-7 | C12H22 | 166.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-03-7
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00008960
InChI Key ZVDBUOGYYYNMQI-UHFFFAOYSA-N
Synonym 1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
PubChem CID 69821
IUPAC Name dodec-1-yne
SMILES CCCCCCCCCCC#C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -19°C
Color Colorless to Yellow
Density 0.7700g/mL
Boiling Point 215°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4330 to 1.435
Linear Formula CH3(CH2)9C∽CH
Quantity 5 mL
Beilstein 01, 261
Specific Gravity 0.77
Formula Weight 166.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dodecyne
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-134-9

RUO – Research Use Only