missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 97%

CAS: 765-03-7 | C12H22 | 166.31 g/mol

Supplier:  Thermo Scientific Chemicals L0297706

 View more versions of this product

Catalog No. AAL0297706

Only null left
Explore available promotions

Description

Description

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.
Specifications

Specifications

1-Dodecyne
-19°C
214°C to 216°C
C12H22
MFCD00008960
1741320
Not miscible in water.
CCCCCCCCCCC#C
166.31
166.31
765-03-7
0.779
79°C (174°F)
1.434
5 g
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
dodec-1-yne
69821
97%

Chemical Identifiers

765-03-7
166.31
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
69821
CCCCCCCCCCC#C
C12H22
MFCD00008960
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
dodec-1-yne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H318-H335

EINECSNumber : 212-134-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only