missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylphosphonic acid, 95%

Catalog No. p-7028848
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2625903 1 g
AAH2625909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2625903 Supplier Thermo Scientific Chemicals Supplier No. H2625903
Only null left

CAS: 5137-70-2 | C12H27O3P | 250.32 g/mol

1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.

Chemical Identifiers

CAS 5137-70-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.32
MDL Number MFCD00015832
InChI Key SVMUEEINWGBIPD-UHFFFAOYSA-N
Synonym n-dodecylphosphonic acid, phosphonic acid, dodecyl, n-dodecanephosphonic acid, dodecane phosphonic acid, 1-dodecanephosphonic acid, n-dodecyl phosphonic acid, dodecyl-phosphonic acid, 1-dodecylphosphonic acid, phosphonic acid, p-dodecyl, n-dodecylphosphonicacid
PubChem CID 78816
IUPAC Name dodecylphosphonic acid
SMILES CCCCCCCCCCCCP(O)(O)=O

Specifications

Melting Point 92°C to 96°C
Quantity 1 g
UN Number UN3261
Solubility Information Sparingly soluble in water.
Formula Weight 250.31
Percent Purity 95%
Chemical Name or Material 1-Dodecylphosphonic acid
Hazard Category H314-H335-H373
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 225-897-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only