missing translation for 'onlineSavingsMsg'
Learn More

(1-Dodecyl)triphenylphosphonium bromide, 98+%

Catalog No. AAA1429530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429530 250 g
AAA1429509 10 g
AAA1429518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429530 Supplier Thermo Scientific Chemicals Supplier No. A1429530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15510-55-1 | C30H40BrP | 511.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15510-55-1
Molecular Formula C30H40BrP
Molecular Weight (g/mol) 511.53
MDL Number MFCD00031546
InChI Key NSIFOGPAKNSGNW-UHFFFAOYSA-M
Synonym dodecyltriphenylphosphonium bromide, unii-2oqv5uaf87, n-dodecyl triphenylphosphonium bromide, phosphonium, dodecyltriphenyl-, bromide, 1-dodecyl triphenylphosphonium bromide, 2oqv5uaf87, dodecyl triphenyl phosphoniumbromide, c12tppbr, n-c12h25pph3br, acmc-1btp1
PubChem CID 84955
ChEBI CHEBI:82638
IUPAC Name dodecyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90°C to 94°C
Quantity 250 g
Beilstein 3581921
Sensitivity Hygroscopic
Formula Weight 511.54
Percent Purity ≥98%
Chemical Name or Material (1-Dodecyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-538-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only