missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecanol, ACS Reagent, ≥98%, Honeywell Riedel-de Haën™
ACS Reagent,≥98.0%
Chemical Identifiers
| CAS | 112-53-8 |
|---|---|
| Molecular Formula | C12H26O |
| Molecular Weight (g/mol) | 186.34 |
| MDL Number | MFCD00004753 |
| InChI Key | LQZZUXJYWNFBMV-UHFFFAOYSA-N |
| Synonym | 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol |
| PubChem CID | 8193 |
| ChEBI | CHEBI:28878 |
| IUPAC Name | dodecan-1-ol |
| SMILES | CCCCCCCCCCCCO |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
| C12H26O | |
| MFCD00004753 | |
| 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
| CHEBI:28878 | |
| CCCCCCCCCCCCO |
Specifications
| 112-53-8 | |
| 0.833g/mL at 25°C | |
| 121°C (249.8°F) | |
| C12H26O | |
| CH3(CH2)11OH | |
| UN3077 | |
| 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
| CCCCCCCCCCCCO | |
| 186.34 | |
| 7.4 (vs air) | |
| 186.33g/mol | |
| 1-Dodecanol |
| 22°C to 26°C | |
| 260°C to 262°C | |
| Conforms to structure | |
| n20/D 1.442 | |
| MFCD00004753 | |
| 1738860 | |
| LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
| dodecan-1-ol | |
| 8193 | |
| CHEBI:28878 | |
| ≥98% |
Safety and Handling
P305 + P351 + P338
DOTInformation : 9/PGIII
RTECSNumber : 203-982-0