missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, 98%

Catalog No. p-7082001
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1222830 250 g
AAA122280B 1000 g
AAA122280I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1222830 Supplier Thermo Scientific Chemicals Supplier No. A1222830
Only null left

CAS: 112-53-8 | C12H26O | 186.34 g/mol

1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

Solubility
Soluble in water (0.004g/L).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Melting Point 24°C to 27°C
Density 0.830
Boiling Point 258°C to 260°C
Flash Point 127°C (260°F)
Assay Percent Range 98%
Refractive Index 1.443
Linear Formula CH3(CH2)11OH
Quantity 250 g
UN Number UN3077
Beilstein 1738860
Merck Index 14,3405
Solubility Information Soluble in water (0.004g/L).
Formula Weight 186.34
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Dodecanol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-982-0
RTECSNumber JR5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only