missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonyl chloride, 97%

Catalog No. AAL0122414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0122414 25 g
AAL0122406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0122414 Supplier Thermo Scientific Chemicals Supplier No. L0122414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10147-40-7 | C12H25ClO2S | 268.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10147-40-7
Molecular Formula C12H25ClO2S
Molecular Weight (g/mol) 268.84
MDL Number MFCD00014728
InChI Key PBULHKIPTBIZHO-UHFFFAOYSA-N
Synonym 1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chloride
PubChem CID 66280
IUPAC Name dodecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 39°C to 42°C
Boiling Point 126°C to 130°C (4 mmHg)
Linear Formula CH3(CH2)11SO2Cl
Quantity 25 g
UN Number UN3261
Beilstein 1778092
Sensitivity Moisture sensitive
Formula Weight 268.84
Percent Purity ≥98%
Chemical Name or Material 1-Dodecanesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-416-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only