accessibility menu, dialog, popup

UserName

H3CSClOO

CAS RN 10147-40-7

IUPAC Name: dodecane-1-sulfonyl chloride
Synonyms: dodecane-1-sulfonyl chloride
Molecular Weight (g/mol): 268.84
Molecular Formula: C12H25ClO2S
InChi Key: PBULHKIPTBIZHO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCS(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)