missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

Catalog No. AC396720250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396720250 25 g Glass bottle
AC396720050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396720250 Supplier Thermo Scientific Chemicals Supplier No. 396720250

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography

Chemical Identifiers

CAS 2386-53-0
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 99.0
Absorbance 0.02 max. (C=0.18 M in hot water) at 230nm, 0.03 max. (C=0.18 M in hot water) at 220nm, 0.05 max. (C=0.18 M in hot water) at 210nm
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C12H25NaO3S
Linear Formula CH3(CH2)11SO3Na
Quantity 25 g
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 272.379
Formula Weight 272.38
Percent Purity ≥99%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecanesulfonic acid, sodium salt, Anhydrous, Ion pair chromatography
Show More Show Less

RUO – Research Use Only