missing translation for 'onlineSavingsMsg'
Learn More

1-(Diphenylmethyl)piperazine, 97%

Catalog No. p-122672
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122935000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC122935000 Supplier Thermo Scientific Chemicals Supplier No. 122935000
Only null left

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 841-77-0
MDL Number MFCD00038379
InChI Key NWVNXDKZIQLBNM-UHFFFAOYSA-N
Synonym 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
PubChem CID 70048
IUPAC Name 1-benzhydrylpiperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 88.0°C to 93.0°C
Color White to Yellow
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C17H20N2
Quantity 500 g
Solubility Information Solubility in water: 0.45g/L (20°C). Other solubilities: 305g/L acetone - >500g/L methanol, >500g/L ethanol - 215g/L 2-propanol, 346g/L dmf - >500g/L methyleenchloride
Molecular Weight (g/mol) 252.36
Formula Weight 252.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-(Diphenylmethyl)piperazine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 212-667-7
RTECSNumber TL6465000
TSCA TSCA

RUO – Research Use Only