missing translation for 'onlineSavingsMsg'
Learn More

1-(Dimethylisopropylsilyl)imidazole 98.0+%, TCI America™

Catalog No. D15961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15961G 1 g
D15965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15961G Supplier TCI America Supplier No. D15961G
Only null left

Chemical Identifiers

CAS 81452-04-2
Molecular Formula C8H16N2Si
Molecular Weight (g/mol) 168.32
MDL Number MFCD00059891
InChI Key OCUUDCWEKWOMFA-UHFFFAOYSA-N
Synonym DMIPS, 1-(Isopropyldimethylsilyl)imidazole
PubChem CID 14325784
IUPAC Name 1-[dimethyl(propan-2-yl)silyl]-1H-imidazole
SMILES CC(C)[Si](C)(C)N1C=CN=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 168.32
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(Dimethylisopropylsilyl)imidazole [Dimethylisopropylsilylating Agent]
TSCA No
Recommended Storage Refrigerator