accessibility menu, dialog, popup

UserName

H3CCH3SiH3CCH3NN

CAS RN 81452-04-2

IUPAC Name: 1-[dimethyl(propan-2-yl)silyl]-1H-imidazole
Synonyms: 1-(isopropyldimethylsilyl)imidazole
Molecular Weight (g/mol): 168.32
Molecular Formula: C8H16N2Si
InChi Key: OCUUDCWEKWOMFA-UHFFFAOYSA-N
SMILES: CC(C)[Si](C)(C)N1C=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results