missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-propanol 98.0+%, TCI America™

Catalog No. p-7126351
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066025ML 25 mL
D0660500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D066025ML Supplier TCI America Supplier No. D066025ML
Only null left

Chemical Identifiers

CAS 108-16-7
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00004532
InChI Key NCXUNZWLEYGQAH-UHFFFAOYSA-N
Synonym 1-dimethylamino-2-propanol, dimepranol, 1-dimethylamino propan-2-ol, n,n-dimethylisopropanolamine, dimethyl 2-hydroxypropyl amine, 2-propanol, 1-dimethylamino, dimethylisopropanolamine, dimepranol inn, n,n-dimethylamino-2-propanol
PubChem CID 37511
IUPAC Name 1-(dimethylamino)propan-2-ol
SMILES CC(CN(C)C)O

Specifications

Melting Point -85°C
Color Yellow
Boiling Point 125°C
Quantity 25 mL
UN Number 2734
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-2-propanol
EINECSNumber (2)-0328&(2)-0353
RTECSNumber UB3150000
TSCA Yes