accessibility menu, dialog, popup

UserName

CAS RN 108-16-7

IUPAC Name: 1-(dimethylamino)propan-2-ol
Synonyms: dimepranol dimethyl isopropanolamine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: NCXUNZWLEYGQAH-UHFFFAOYNA-N
SMILES: CC(O)CN(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)