missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dimethylamino-2-propanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -85°C |
| Color | Yellow |
| Boiling Point | 125°C |
| Quantity | 500 mL |
| UN Number | 2734 |
| Formula Weight | 103.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Dimethylamino-2-propanol |
Safety and Handling
| EINECSNumber | (2)-0328&(2)-0353 |
| RTECSNumber | UB3150000 |
| TSCA | Yes |