missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-propanol, 99%

Catalog No. p-7027462
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2423430 250 g
AAB242340B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2423430 Supplier Thermo Scientific Chemicals Supplier No. B2423430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-16-7 | C5H13NO | 103.165 g/mol

1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Chemical Identifiers

CAS 108-16-7
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00004532
InChI Key NCXUNZWLEYGQAH-UHFFFAOYSA-N
Synonym 1-dimethylamino-2-propanol, dimepranol, 1-dimethylamino propan-2-ol, n,n-dimethylisopropanolamine, dimethyl 2-hydroxypropyl amine, 2-propanol, 1-dimethylamino, dimethylisopropanolamine, dimepranol inn, n,n-dimethylamino-2-propanol
PubChem CID 37511
IUPAC Name 1-(dimethylamino)propan-2-ol
SMILES CC(CN(C)C)O

Specifications

Melting Point -85°C
Density 0.85
pH 11
Boiling Point 123°C to 126°C
Flash Point 35°C (95°F)
Odor Amine-like
Refractive Index 1.4195
Quantity 250 g
UN Number UN2734
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 1-Dimethylamino-2-propanol
Show More Show Less
Hazard Category H226-H302-H314
Hazard Statement GHS H Statement
H314-H226-H302
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 203-556-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only