missing translation for 'onlineSavingsMsg'
Learn More

1-Diazo-2-naphthol-4-sulfonic acid, 76%, Thermo Scientific™

$42.01 - $113.34

Chemical Identifiers

CAS 887-76-3
Molecular Formula C10H6N2O4S
MDL Number MFCD00012409
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6080514
View Documents
Thermo Scientific Chemicals
H6080514
25 g
Each for $42.01
Only null left
 
AAH6080522
View Documents
Thermo Scientific Chemicals
H6080522
100 g
Each for $113.34
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

887-76-3
C10H6N2O4S
MFCD00012409

Specifications

887-76-3
C10H6N2O4S
25 g
250.23
1-Diazo-2-naphthol-4-sulfonic acid
160°C (decomposition)
MFCD00012409
UN2925
76%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 212-958-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'