Learn More
1-Diazo-2-naphthol-4-sulfonic acid, 76%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6080522
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1-Diazo-2-naphthol-4-sulfonic acid | |
| 160°C (decomposition) | |
| MFCD00012409 | |
| UN2925 | |
| 76% |
| 887-76-3 | |
| C10H6N2O4S | |
| 100 g | |
| 250.23 |
Chemical Identifiers
| 887-76-3 | |
| C10H6N2O4S | |
| MFCD00012409 |
Safety and Handling
GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 212-958-9
TSCA : Yes
Recommended Storage : Ambient temperatures