missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxynojirimycin hydrochloride, 98%

Catalog No. p-7043238
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66458MA 10 mg
AAJ66458MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66458MA Supplier Thermo Scientific Chemicals Supplier No. J66458MA
Only null left

CAS: 73285-50-4 | C6H14ClNO4 | 199.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73285-50-4
Molecular Formula C6H14ClNO4
Molecular Weight (g/mol) 199.631
MDL Number MFCD00133247
InChI Key ZJIHMALTJRDNQI-VFQQELCFSA-N
Synonym duvoglustat hcl, duvoglustat hydrochloride, deoxynojirimycin hydrochloride, 1-deoxynojirimycin hydrochloride, unii-0rn23c42qr, duvoglustat hydrochloride usan, +-1-deoxynojirimycin hydrochloride, moranoline hydrochloride, 1-deoxy-l-altronojirimycin hydrochloride
PubChem CID 13018787
IUPAC Name (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES C1C(C(C(C(N1)CO)O)O)O.Cl

Specifications

Color White
Quantity 10 mg
Beilstein 3563225
Solubility Information Soluble in water at 20mg/ml
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Deoxynojirimycin hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only