accessibility menu, dialog, popup

UserName

CAS RN 73285-50-4

IUPAC Name: 2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
Synonyms: 1 deoxynojirimycin hydrochloride
Molecular Weight (g/mol): 199.63
Molecular Formula: C6H14ClNO4
InChi Key: ZJIHMALTJRDNQI-UHFFFAOYNA-N
SMILES: Cl.OCC1NCC(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results