missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 1-Decyne | |
| Yellow | |
| C10H18 | |
| 5 mL | |
| 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y | |
| CCCCCCCCC#C | |
| 138.254 | |
| CHEBI:87322 | |
| ≥95.0% (GC) |
| 764-93-2 | |
| 174°C | |
| MFCD00009576 | |
| 3295 | |
| ILLHQJIJCRNRCJ-UHFFFAOYSA-N | |
| dec-1-yne | |
| 12997 | |
| 138.25 | |
| Liquid |
Chemical Identifiers
| 764-93-2 | |
| 138.254 | |
| ILLHQJIJCRNRCJ-UHFFFAOYSA-N | |
| 12997 | |
| dec-1-yne |
| C10H18 | |
| MFCD00009576 | |
| 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y | |
| CHEBI:87322 | |
| CCCCCCCCC#C |
Safety and Handling
TSCA : Yes