missing translation for 'onlineSavingsMsg'
Learn More

1-Decyne 95.0+%, TCI America™

Catalog No. p-7125996
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
D00375ML 5 mL
D003725ML 25 mL
2 options

Catalog No. D00375ML

Supplier: TCI America D00375ML

Only null left

Chemical Identifiers

CAS 764-93-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00009576
InChI Key ILLHQJIJCRNRCJ-UHFFFAOYSA-N
Synonym 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
PubChem CID 12997
ChEBI CHEBI:87322
IUPAC Name dec-1-yne
SMILES CCCCCCCCC#C

Specifications

Color Yellow
Boiling Point 174°C
Quantity 5 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Decyne