missing translation for 'onlineSavingsMsg'
Learn More

1-Decylphosphonic acid, 98%

Catalog No. AAL1201906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1201906 5 g
AAL1201903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1201906 Supplier Thermo Scientific Chemicals Supplier No. L1201906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6874-60-8 | C10H23O3P | 222.265 g/mol

1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6874-60-8
Molecular Formula C10H23O3P
Molecular Weight (g/mol) 222.265
MDL Number MFCD00015830
InChI Key DZQISOJKASMITI-UHFFFAOYSA-N
Synonym decanephosphonic acid, phosphonic acid, decyl, 1-decanephosphonic acid, decyl-phosphonic acid, 1-decylphosphonic acid, n-decylphosphonic acid, phosphonic acid, p-decyl, decylphosphonicacid, n-decanephosphonic acid, acmc-1cuhv
PubChem CID 81309
IUPAC Name decylphosphonic acid
SMILES CCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 100°C to 101°C
Quantity 5 g
UN Number UN3261
Beilstein 1771572
Formula Weight 222.27
Percent Purity 98%
Chemical Name or Material 1-Decylphosphonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 229-975-2
RTECSNumber SZ7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only