missing translation for 'onlineSavingsMsg'
Learn More

1-Decyl-3-methylimidazolium Bromide 98.0+%, TCI America™

Catalog No. p-7128747
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53505G 5 g
D535025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53505G Supplier TCI America Supplier No. D53505G

Chemical Identifiers

CAS 188589-32-4
Molecular Formula C14H27BrN2
Molecular Weight (g/mol) 303.29
MDL Number MFCD09038832
InChI Key HOISBTKPPVRFDS-UHFFFAOYSA-M
Synonym 1-Decyl-3-methyl-1H-imidazol-3-ium Bromide
PubChem CID 22078297
IUPAC Name 1-decyl-3-methyl-1H-imidazol-3-ium bromide
SMILES [Br-].CCCCCCCCCCN1C=C[N+](C)=C1

Specifications

Quantity 5 g
Formula Weight 303.29
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Decyl-3-methylimidazolium Bromide