missing translation for 'onlineSavingsMsg'
Learn More

1-Decene 95.0+%, TCI America™

Catalog No. p-7125991
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D002825ML 25 mL
D0028500ML 500 mL
2 options

Catalog No. D002825ML

Supplier: TCI America D002825ML

Only null left

Chemical Identifiers

CAS 872-05-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009577
InChI Key AFFLGGQVNFXPEV-UHFFFAOYSA-N
Synonym 1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
PubChem CID 13381
ChEBI CHEBI:87315
IUPAC Name dec-1-ene
SMILES CCCCCCCCC=C

Specifications

Melting Point -66°C
Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Decene
EINECSNumber (2)-0027
RTECSNumber HE2071401
TSCA Yes