accessibility menu, dialog, popup

UserName

H3CCH2

CAS RN 872-05-9

IUPAC Name: dec-1-ene
Synonyms: decene decene (mixture of isomers) hydrogenated polydecene α-olefins (petroleum)
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: AFFLGGQVNFXPEV-UHFFFAOYSA-N
SMILES: CCCCCCCCC=C
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)