missing translation for 'onlineSavingsMsg'
Learn More

1-Decene 95.0+%, TCI America™

Catalog No. D002825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D002825ML 25 mL
D0028500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D002825ML Supplier TCI America Supplier No. D002825ML

Chemical Identifiers

CAS 872-05-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009577
InChI Key AFFLGGQVNFXPEV-UHFFFAOYSA-N
Synonym 1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
PubChem CID 13381
ChEBI CHEBI:87315
IUPAC Name dec-1-ene
SMILES CCCCCCCCC=C

Specifications

Melting Point -66°C
Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Decene
EINECSNumber (2)-0027
RTECSNumber HE2071401
TSCA Yes