missing translation for 'onlineSavingsMsg'
Learn More

1-Decene, ca. 95%

Catalog No. p-4258922
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111910010 1 L Glass bottle
AC111910025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111910010 Supplier Thermo Scientific Chemicals Supplier No. 111910010
Only null left

CAS: 872-05-9 | C10H20 | 140.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 872-05-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
InChI Key AFFLGGQVNFXPEV-UHFFFAOYSA-N
Synonym 1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
PubChem CID 13381
ChEBI CHEBI:87315
IUPAC Name dec-1-ene
SMILES CCCCCCCCC=C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -66.0°C
Color Colorless
Density 0.7410g/mL
Boiling Point 169.0°C to 171.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH2
Refractive Index 1.4200 to 1.4220
Quantity 1 L
Beilstein 01,III,858
Specific Gravity 0.741
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Viscosity 1.1 mm2/s (20°C)
Formula Weight 140.27
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material 1-Decene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 212-819-2

RUO – Research Use Only