missing translation for 'onlineSavingsMsg'
Learn More

1-Decanol 98.0+%, TCI America™

Catalog No. D003125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003125ML 25 mL
D0031500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D003125ML Supplier TCI America Supplier No. D003125ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-30-1
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00004747
InChI Key MWKFXSUHUHTGQN-UHFFFAOYSA-N
Synonym 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
PubChem CID 8174
ChEBI CHEBI:28903
IUPAC Name decan-1-ol
SMILES CCCCCCCCCCO

Specifications

Color Colorless
Boiling Point 233°C
Quantity 25 mL
UN Number 3082
Formula Weight 158.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Decanol
EINECSNumber (2)-0217
RTECSNumber HE4375000
TSCA Yes