missing translation for 'onlineSavingsMsg'
Learn More

1-Decanol, MilliporeSigma™ Supelco™

Catalog No. 111020639
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020639 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020639 Supplier Merck Emd Millipore Supplier No. 442539
Only null left

1-Decanol can be obtained via decomposition of decyl hydroperoxide or dehydrogenation of 1-decene, followed by hydrolysis.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Ignition Point 550°F
CAS 112-30-1
Vapor Pressure 8.25 mmHg (100°C)
Vapor Density 4.5 (vs. air)
Density 0.829 g/mL (at 25°C)
Flash Point 82°C
Physical Form Neat
Linear Formula CH3(CH2)9 OH
MDL Number MFCD00004747
Melting Point 5°C to 7°C
Boiling Point 231°C
Synonym n-Decyl alcohol; Alcohol C10
Solubility Slightly soluble in water
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 158.28
Grade Analytical Standard
Show More Show Less