missing translation for 'onlineSavingsMsg'
Learn More

1-Decanol, 98+%

Catalog No. aaa172880f
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA172880F 2500 mL
AAA17288AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA172880F Supplier Thermo Scientific Chemicals Supplier No. A172880F
Only null left

CAS: 112-30-1 | C10H22O | 158.285 g/mol

1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 112-30-1
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00004747
InChI Key MWKFXSUHUHTGQN-UHFFFAOYSA-N
Synonym 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
PubChem CID 8174
ChEBI CHEBI:28903
IUPAC Name decan-1-ol
SMILES CCCCCCCCCCO

Specifications

Melting Point 6°C
Density 0.828
Boiling Point 230°C to 232°C
Flash Point 82°C (179°F)
Odor Sweet
Assay Percent Range >98%
Refractive Index 1.437
Linear Formula CH3(CH2)8CH2OH
Quantity 2500 mL
UN Number UN3082
Beilstein 1735221
Merck Index 14,2855
Solubility Information Soluble in alcohol,ether; Insoluble in water;Miscible with carbon tetrachloride,ethanol,ether,acetone,benzene,chloroform and glacial acetic acid; Immiscible with water.
Formula Weight 158.29
Percent Purity ≥98%
Chemical Name or Material 1-Decanol
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H332-H315-H319-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-956-9
RTECSNumber HE4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only