missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic Acid Sodium Salt, Approx. 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D200425GM

 View more versions of this product

Catalog No. 18608098


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

13419-61-9
Amber Glass Bottle
∼99%
1
25 g
Reagent

Chemical Identifiers

13419-61-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.