missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic Acid Sodium Salt, Approx. 99%, Spectrum™ Chemical

Catalog No. p-7165482
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18608097 5 g Amber Glass Bottle
18608098 25 g Amber Glass Bottle
18608099 100 g Amber Glass Bottle
18608100 1 kg Poly Pail
18608101 5 kg Poly Pail
5 options

Catalog No. 18608097

Supplier: Spectrum Chemical Mfg Cor D20045GM

Only null left

Chemical Identifiers

CAS 13419-61-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity ∼99%
Grade Reagent

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.