missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropylethanol 98.0+%, TCI America™

Supplier:  TCI America C098425ML

 View more versions of this product

Catalog No. C098425ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Cyclopropylethanol
Yellow
C5H10O
25 mL
cyclopropylmethylcarbinol, cyclopropyl methyl carbinol, methylcyclopropylcarbinol, 1-cyclopropylethan-1-ol, cyclopropylethanol, cyclopropanemethanol, .alpha.-methyl, alpha-methylcyclopropanemethanol, cyclopropanemethanol, a-methyl, cyclopropyl ethanol, cyclopropanemethanol, alpha-methyl
CC(C1CC1)O
86.134
86.13
Liquid
765-42-4
122°C
MFCD00001300
1987
DKKVKJZXOBFLRY-UHFFFAOYSA-N
1-cyclopropylethanol
79104
≥98.0% (GC)

Chemical Identifiers

765-42-4
86.134
DKKVKJZXOBFLRY-UHFFFAOYSA-N
79104
CC(C1CC1)O
C5H10O
MFCD00001300
cyclopropylmethylcarbinol, cyclopropyl methyl carbinol, methylcyclopropylcarbinol, 1-cyclopropylethan-1-ol, cyclopropylethanol, cyclopropanemethanol, .alpha.-methyl, alpha-methylcyclopropanemethanol, cyclopropanemethanol, a-methyl, cyclopropyl ethanol, cyclopropanemethanol, alpha-methyl
1-cyclopropylethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions