missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropylethanol 98.0+%, TCI America™

Catalog No. C09845ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09845ML 5 mL
C098425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09845ML Supplier TCI America Supplier No. C09845ML

Chemical Identifiers

CAS 765-42-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001300
InChI Key DKKVKJZXOBFLRY-UHFFFAOYSA-N
Synonym cyclopropylmethylcarbinol, cyclopropyl methyl carbinol, methylcyclopropylcarbinol, 1-cyclopropylethan-1-ol, cyclopropylethanol, cyclopropanemethanol, .alpha.-methyl, alpha-methylcyclopropanemethanol, cyclopropanemethanol, a-methyl, cyclopropyl ethanol, cyclopropanemethanol, alpha-methyl
PubChem CID 79104
IUPAC Name 1-cyclopropylethanol
SMILES CC(C1CC1)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclopropylethanol