missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%
CAS: 1021298-67-8 | C8H15ClN2O | 190.67 g/mol
Supplier: Thermo Scientific Chemicals H6624206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-(Cyclopropylcarbonyl)piperazine hydrochloride | |
| 175°C to 179°C | |
| C8H15ClN2O | |
| 5 g | |
| 1-cyclopropylcarbonyl piperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hydrochloride, 1-cyclopropanecarbonylpiperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hcl, cyclopropylcarboxylic acid-1-piperazineamide hydrochloride, cyclopropyl piperazinyl ketone, chloride, pubchem19546, 1-cyclopropylcarbonyl piperazine hcl, 1-cyclopropylcarbonyl-piperazine hcl, 1-cyclopropylcarbonyl piperazine hcl salt | |
| Cl.O=C(C1CC1)N1CCNCC1 | |
| 190.67 | |
| 190.67 | |
| Solid |
| 1021298-67-8 | |
| White | |
| MFCD03428576 | |
| 18312533 | |
| WIHDAPMHNYYTOA-UHFFFAOYSA-N | |
| cyclopropyl(piperazin-1-yl)methanone;hydrochloride | |
| 20847955 | |
| 97% |
Chemical Identifiers
| 1021298-67-8 | |
| 190.67 | |
| WIHDAPMHNYYTOA-UHFFFAOYSA-N | |
| 20847955 | |
| Cl.O=C(C1CC1)N1CCNCC1 |
| C8H15ClN2O | |
| MFCD03428576 | |
| 1-cyclopropylcarbonyl piperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hydrochloride, 1-cyclopropanecarbonylpiperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hcl, cyclopropylcarboxylic acid-1-piperazineamide hydrochloride, cyclopropyl piperazinyl ketone, chloride, pubchem19546, 1-cyclopropylcarbonyl piperazine hcl, 1-cyclopropylcarbonyl-piperazine hcl, 1-cyclopropylcarbonyl piperazine hcl salt | |
| cyclopropyl(piperazin-1-yl)methanone;hydrochloride |
RUO – Research Use Only