missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%

Catalog No. p-7056983
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66242MD 250 mg
AAH6624203 1 g
AAH6624206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66242MD Supplier Thermo Scientific Chemicals Supplier No. H66242MD
Only null left

CAS: 1021298-67-8 | C8H15ClN2O | 190.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1021298-67-8
Molecular Formula C8H15ClN2O
Molecular Weight (g/mol) 190.67
MDL Number MFCD03428576
InChI Key WIHDAPMHNYYTOA-UHFFFAOYSA-N
Synonym 1-cyclopropylcarbonyl piperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hydrochloride, 1-cyclopropanecarbonylpiperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hcl, cyclopropylcarboxylic acid-1-piperazineamide hydrochloride, cyclopropyl piperazinyl ketone, chloride, pubchem19546, 1-cyclopropylcarbonyl piperazine hcl, 1-cyclopropylcarbonyl-piperazine hcl, 1-cyclopropylcarbonyl piperazine hcl salt
PubChem CID 20847955
IUPAC Name cyclopropyl(piperazin-1-yl)methanone;hydrochloride
SMILES Cl.O=C(C1CC1)N1CCNCC1

Specifications

Melting Point 175°C to 179°C
Color White
Quantity 250 mg
Beilstein 18312533
Formula Weight 190.67
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine hydrochloride

RUO – Research Use Only