missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenylboronic Acid (contains varying amounts of Anhydride), TCI America™

$121.28 - $121.28

Chemical Identifiers

CAS 850036-28-1
Molecular Formula C5H9BO2
Molecular Weight (g/mol) 111.935
MDL Number MFCD02179496
InChI Key UZBHNSVUMGIKLU-UHFFFAOYSA-N
PubChem CID 3861343
IUPAC Name cyclopenten-1-ylboronic acid
SMILES B(C1=CCCC1)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C28931G
View Documents
TCI America
C28931G
1 g
Each for $121.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

850036-28-1
111.935
UZBHNSVUMGIKLU-UHFFFAOYSA-N
cyclopenten-1-ylboronic acid
C5H9BO2
MFCD02179496
3861343
B(C1=CCCC1)(O)O

Specifications

850036-28-1
C5H9BO2
1 g
UZBHNSVUMGIKLU-UHFFFAOYSA-N
cyclopenten-1-ylboronic acid
3861343
Crystalline Powder
140°C
MFCD02179496
cyclopent-1-en-1-ylboronic acid, 1-cyclopentenylboronic acid, cyclopentenylboronic acid, cyclopent-1-en-1-yl boronic acid, cyclopentene-1-boronic acid, cyclopenten-1-yl boronic acid, cyclopent-1-ene-1-boronic acid, boronic acid, 1-cyclopenten-1-yl, cyclopenteneboronic acid, acmc-209pzb
B(C1=CCCC1)(O)O
111.935
111.94
1-Cyclopentenylboronic Acid (contains varying amounts of Anhydride)
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents