missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-2-buten-1-ol (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. C15675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15675G Supplier TCI America Supplier No. C15675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18736-82-8
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00060819
InChI Key WZIIBXRIIUAFOT-QHHAFSJGSA-N
Synonym 1r,2z-1-cyclohexylbut-2-en-1-ol
PubChem CID 5358342
IUPAC Name (E)-1-cyclohexylbut-2-en-1-ol
SMILES CC=CC(C1CCCCC1)O

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexyl-2-buten-1-ol (cis- and trans- mixture)