accessibility menu, dialog, popup

UserName

CAS RN 18736-82-8

IUPAC Name: (1R,2Z)-1-cyclohexylbut-2-en-1-ol
Synonyms: (1R,2Z)-1-cyclohexylbut-2-en-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WZIIBXRIIUAFOT-RILPQGNRSA-N
SMILES: C\C=C/[C@H](O)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results