missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-1-pentanol 95.0+%, TCI America™

Catalog No. c15695g
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
C15695G 5 g
1 options

Catalog No. C15695G

Supplier: TCI America C15695G

Only null left

Chemical Identifiers

CAS 7338-43-4
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00045483
InChI Key PKXSPCMDZCKLCI-UHFFFAOYSA-N
Synonym 1-cyclohexyl-1-pentanol, cyclohexylpentanol-1, 1-cyclohexyl-pentan-1-ol, acmc-20aj12, n-butyl cyclohexyl carbinol, 1-hydroxypentyl cyclohexane, 1-cyclohexyl pentanol-1, pkxspcmdzcklci-uhfffaoysa, --1-cyclohexyl-1-pentanol
PubChem CID 145270
IUPAC Name 1-cyclohexylpentan-1-ol
SMILES CCCCC(C1CCCCC1)O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 170.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexyl-1-pentanol