accessibility menu, dialog, popup

UserName

H3C(S)OH

CAS RN 7338-43-4

IUPAC Name: (1S)-1-phenylpentan-1-ol
Synonyms: (1S)-1-phenylpentan-1-ol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: OVGORFFCBUIFIA-NSHDSACASA-N
SMILES: CCCC[C@H](O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results