missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1,2-dicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. C17015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17015G 5 g
C170125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17015G Supplier TCI America Supplier No. C17015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione
PubChem CID 12475
IUPAC Name 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 72°C
Color White
Quantity 5 g
UN Number 2698
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclohexene-1,2-dicarboxylic Anhydride
EINECSNumber (3)-3435
TSCA Yes