Learn More
1-Cyclohexene-1,2-dicarboxylic anhydride, 97+%
CAS: 2426-02-0 | C8H8O3 | 152.15 g/mol
$450.41 - $450.41
Chemical Identifiers
| CAS | 2426-02-0 |
|---|---|
| Molecular Formula | C8H8O3 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00005917 |
| InChI Key | HMMBJOWWRLZEMI-UHFFFAOYSA-N |
| Synonym | 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione |
| PubChem CID | 12475 |
| IUPAC Name | 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione |
| SMILES | O=C1OC(=O)C2=C1CCCC2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5692922
|
Thermo Scientific Chemicals
H5692922 |
100 g |
Each for $450.41
|
|
|||||
Description
It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.
Solubility
Reacts with water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 2426-02-0 | |
| 152.15 | |
| HMMBJOWWRLZEMI-UHFFFAOYSA-N | |
| 12475 | |
| O=C1OC(=O)C2=C1CCCC2 |
| C8H8O3 | |
| MFCD00005917 | |
| 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione | |
| 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione |
Specifications
| 2426-02-0 | |
| C8H8O3 | |
| 100 g | |
| 136335 | |
| Reacts with water. | |
| O=C1OC(=O)C2=C1CCCC2 | |
| 152.15 | |
| 152.15 | |
| 1-Cyclohexene-1,2-dicarboxylic anhydride |
| 69°C to 73°C | |
| MFCD00005917 | |
| UN2698 | |
| 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione | |
| HMMBJOWWRLZEMI-UHFFFAOYSA-N | |
| 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione | |
| 12475 | |
| ≥97% |
Safety and Handling
GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
H317-H318-H334
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TETRAHYDROPHTHALIC ANHYDRIDES
EINECSNumber : 219-374-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only