missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyanoacetyl)pyrrolidine, 98+%

CAS: 14227-95-3 | C7H10N2O | 138.17 g/mol

$55.44 - $156.40

Chemical Identifiers

CAS 14227-95-3
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00020838
InChI Key VEUDVNNBYYRZBV-UHFFFAOYSA-N
Synonym 1-cyanoacetyl pyrrolidine, 3-oxo-3-pyrrolidin-1-yl propanenitrile, 1-cyanoacetylpyrrolidine, 3-oxo-3-1-pyrrolidinyl propanenitrile, pyrrolidine, 1-cyanoacetyl, 1-pyrrolidinepropanenitrile,b-oxo, 3-oxo-3-pyrrolidinylpropanenitrile, n-cyanoacetylpyrrolidine, acmc-20ak5f, oxopyrrolidinylpropanenitrile
PubChem CID 84272
IUPAC Name 3-oxo-3-pyrrolidin-1-ylpropanenitrile
SMILES O=C(CC#N)N1CCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1252303
View Documents
Thermo Scientific Chemicals
A1252303
1 g
Each for $55.44
Only null left
 
AAA1252306
View Documents
Thermo Scientific Chemicals
A1252306
5 g
Each for $156.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14227-95-3
138.17
VEUDVNNBYYRZBV-UHFFFAOYSA-N
84272
O=C(CC#N)N1CCCC1
C7H10N2O
MFCD00020838
1-cyanoacetyl pyrrolidine, 3-oxo-3-pyrrolidin-1-yl propanenitrile, 1-cyanoacetylpyrrolidine, 3-oxo-3-1-pyrrolidinyl propanenitrile, pyrrolidine, 1-cyanoacetyl, 1-pyrrolidinepropanenitrile,b-oxo, 3-oxo-3-pyrrolidinylpropanenitrile, n-cyanoacetylpyrrolidine, acmc-20ak5f, oxopyrrolidinylpropanenitrile
3-oxo-3-pyrrolidin-1-ylpropanenitrile

Specifications

14227-95-3
Odorless
MFCD00020838
637151
VEUDVNNBYYRZBV-UHFFFAOYSA-N
3-oxo-3-pyrrolidin-1-ylpropanenitrile
84272
≥98%
71°C to 74°C
C7H10N2O
1 g
1-cyanoacetyl pyrrolidine, 3-oxo-3-pyrrolidin-1-yl propanenitrile, 1-cyanoacetylpyrrolidine, 3-oxo-3-1-pyrrolidinyl propanenitrile, pyrrolidine, 1-cyanoacetyl, 1-pyrrolidinepropanenitrile,b-oxo, 3-oxo-3-pyrrolidinylpropanenitrile, n-cyanoacetylpyrrolidine, acmc-20ak5f, oxopyrrolidinylpropanenitrile
O=C(CC#N)N1CCCC1
138.17
138.17
1-(Cyanoacetyl)pyrrolidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 238-096-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only