CAS RN 14227-95-3
IUPAC Name:
3-oxo-3-(pyrrolidin-1-yl)propanenitrile
Synonyms:
3-oxo-3-(pyrrolidin-1-yl)propanenitrile
Molecular Weight (g/mol):
138.17
Molecular Formula:
C7H10N2O
InChi Key:
VEUDVNNBYYRZBV-UHFFFAOYSA-N
SMILES:
O=C(CC#N)N1CCCC1
1
–
1
of
1
results
1
Promotions Available
1-(Cyanoacetyl)pyrrolidine, 98+%
1-(Cyanoacetyl)pyrrolidine, 98+%
CAS: 14227-95-3 Molecular Formula: C7H10N2O Molecular Weight (g/mol): 138.17 MDL Number: MFCD00020838 InChI Key: VEUDVNNBYYRZBV-UHFFFAOYSA-N Synonym: 1-cyanoacetyl pyrrolidine,3-oxo-3-pyrrolidin-1-yl propanenitrile,1-cyanoacetylpyrrolidine,3-oxo-3-1-pyrrolidinyl propanenitrile,pyrrolidine, 1-cyanoacetyl,1-pyrrolidinepropanenitrile,b-oxo,3-oxo-3-pyrrolidinylpropanenitrile,n-cyanoacetylpyrrolidine,acmc-20ak5f,oxopyrrolidinylpropanenitrile PubChem CID: 84272 IUPAC Name: 3-oxo-3-pyrrolidin-1-ylpropanenitrile SMILES: O=C(CC#N)N1CCCC1